missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ F(ab')2-Goat anti-Human IgG (H+L) Secondary Antibody, PE

F(ab')2-Goat Polyclonal Secondary Antibody

Brand:  Invitrogen™ PA184341

Product Code. 11509150

  • £512.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

F(ab')2 fragments were prepared by pepsin digestion. PA1-84341 detects IgG in human samples. This antibody does not cross react with goat, mouse, rat, or equine serum proteins. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Goat F(ab')2 anti Human IgG antibody recognizes human IgG and demonstrates Goat F(ab')2 anti Human IgG antibody does not cross-react with Goat IgG F(c) or mouse, rat, bovine and horse serum proteins, as determined by immunoelectrophoresis.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Human IgG (H+L)
Polyclonal
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
Human IgG.
1 mL
Secondary
4°C, store in dark, DO NOT FREEZE!
Lyophilized
Flow Cytometry
PE
Goat
Affinity chromatography
RUO
Human
Secondary Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • F(ab')2-Goat anti-Human IgG (H+L) Secondary Antibody, PE

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sodium azide

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.