missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F(ab')2-Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, HRP
F(ab')2-Donkey Polyclonal Secondary Antibody
Brand: Invitrogen™ A24506
This item is not returnable.
View return policy
Description
F(ab')2-Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody for ELISA, IHC, Western Blot
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Mouse IgG (H+L) Highly Cross-Adsorbed | |
| Polyclonal | |
| HRP | |
| Donkey | |
| Affinity Chromatography | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA and 0.1% ProClin 150; pH 7.2 | |
| Gamma Immunoglobins Heavy and Light chains. | |
| 500 μg | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction