missing translation for 'onlineSavingsMsg'
Learn More

Sartorius F-10 (HAM) Nutrient Mixture with L-Glutamine

Product Code. 31908480 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31908480 500 mL 500mL
31908348 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31908480 Supplier Sartorius Supplier No. 010901A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ham's F-10 Nutrient Mixture (F-10) was designed for serum-free growth of Chinese Hamster Ovary (CHO) cells.

  • F-10 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including primary rat astrocytes and epithelial cells.

Specifications

Form Liquid
Product Type Ham's F-10 Cell Culture Media
With Additives Glutamine, Sodium Bicarbonate
Without Additives None
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.