missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ezrin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26056-1-AP-20UL
This item is not returnable.
View return policy
Description
The cytoplasmic peripheral membrane protein encoded by this gene functions as a protein-tyrosine kinase substrate in microvilli. As a member of the ERM protein family, this protein serves as an intermediate between the plasma membrane and the actin cytoskeleton. This protein plays a key role in cell surface structure adhesion, migration and organization, and it has been implicated in various human cancers. A pseudogene located on chromosome 3 has been identified for this gene. Alternatively spliced variants have also been described for this gene.Specifications
| Ezrin | |
| Polyclonal | |
| Unconjugated | |
| EZR | |
| CVIL, CVL, Cytovillin, EZR, Ezrin, p81, VIL2, Villin 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22350, 7430 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P15311, P26040 | |
| EZR | |
| Ezrin Fusion Protein Ag23530 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction