missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ezrin Antibody (7F11), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Ezrin |
|---|---|
| Clone | 7F11 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043706
|
Novus Biologicals
NBP3-26596-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043619
|
Novus Biologicals
NBP3-26596-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ezrin Monoclonal antibody specifically detects Ezrin in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| Ezrin | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2MGC1584, villin 2 (ezrin), villin-2 | |
| A synthesized peptide derived from Human Ezrin [UniProt P15311] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 7F11 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Phospho Specific, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 7430 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title