missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ezrin Antibody (2086A), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB7239MFV500
This item is not returnable.
View return policy
Description
Ezrin Monoclonal antibody specifically detects Ezrin in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
Specifications
| Ezrin | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Phospho Specific, Signal Transduction | |
| 7430 | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
| 2086A | |
| Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2MGC1584, villin 2 (ezrin), villin-2 | |
| E. coli-derived recombinant human Ezrin, Lys438-Arg562, Accession # P15311 | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction