Learn More
Toffeln™ Ezi-Klog Shoes with Adjustable Heel Strap, Black
Click to view available options
Size:
3
4
5
6
7
8
9
10
11
12
Description
- Adjustable heel strap
- Lightweight
- Closed-in heel
- Shock absorbing Anti-static heel
- Side vents
- Exceeds EN20347 SRC
Specifications
Specifications
| Color | Black |
| Material | E-tech |
| Size | 11 |
| Style | FW516 |