missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EZH2 Recombinant Rabbit Monoclonal Antibody (2C7)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA549830
This item is not returnable.
View return policy
Description
EZH2 Recombinant Monoclonal Antibody for IHC (P), ELISA
EZH2 (Histone-lysine N-methyltransferase) is a member of the Polycomb-group (PcG) family. PcG family members form multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. This protein associates with the embryonic ectoderm development protein, the VAV1 oncoprotein, and the X-linked nuclear protein. EZH2 may play a role in the hematopoietic and central nervous systems. Mutations affecting EZH2 can cause Weaver syndrome (WVS).
Specifications
| EZH2 | |
| Recombinant Monoclonal | |
| 0.65 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15910 | |
| EZH2 | |
| A synthesized peptide derived from human KMT6/EZH2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 2C7 | |
| Unconjugated | |
| EZH2 | |
| an; enhancer of zeste 2 polycomb repressive complex 2 subunit; enhancer of zeste homolog 2; enhancer of zeste homolog 2 (Drosophila); ENX 1; ENX1; Enx-1; Enx1h; EZH 2; EZH1; EZH2; EZH-2; EZH2b; histone-lysine N-methyltransferase EZH2; KMT6; KMT6A; lysine N-methyltransferase 6; mKIAA4065; OTTHUMP00000213774; OTTHUMP00000213778; OTTHUMP00000213779; OTTHUMP00000213780; WVS; WVS2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2146 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction