missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EZH2 Recombinant Rabbit Monoclonal Antibody (23GB835), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | EZH2 |
|---|---|
| Clone | 23GB835 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089512
![]() |
Invitrogen™
MA554232 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087904
|
Invitrogen™
MA554233 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EZH2 Recombinant Monoclonal antibody specifically detects EZH2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| EZH2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q15910, Q61188 | |
| 14056, 2146, 312299 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB835 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| an; enhancer of zeste 2 polycomb repressive complex 2 subunit; enhancer of zeste homolog 2; enhancer of zeste homolog 2 (Drosophila); ENX 1; ENX1; Enx-1; Enx1h; EZH 2; EZH1; EZH2; EZH-2; EZH2b; histone-lysine N-methyltransferase EZH2; KMT6; KMT6A; lysine N-methyltransferase 6; mKIAA4065; OTTHUMP00000213774; OTTHUMP00000213778; OTTHUMP00000213779; OTTHUMP00000213780; WVS; WVS2 | |
| A synthetic peptide of human KMT6/EZH2 (1-55AA). | |
| Primary | |
| -20°C | |
| EZH2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title