missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EZH2 Monoclonal Antibody (144CT2.1.1.5)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA518108

Product Code. 15367374

  • £487.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is predicted to react with non-human primate based on sequence homology.

EZH2 (Histone-lysine N-methyltransferase) is a member of the Polycomb-group (PcG) family. PcG family members form multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. This protein associates with the embryonic ectoderm development protein, the VAV1 oncoprotein, and the X-linked nuclear protein. EZH2 may play a role in the hematopoietic and central nervous systems. Mutations affecting EZH2 can cause Weaver syndrome (WVS).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EZH2
Monoclonal
0.48 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q15910
EZH2
EZH2 recombinant protein.
400 μL
Primary
Human
Antibody
IgG1 κ
ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
144CT2.1.1.5
Unconjugated
EZH2
an; enhancer of zeste 2 polycomb repressive complex 2 subunit; enhancer of zeste homolog 2; enhancer of zeste homolog 2 (Drosophila); ENX 1; ENX1; Enx-1; Enx1h; EZH 2; EZH1; EZH2; EZH-2; EZH2b; histone-lysine N-methyltransferase EZH2; KMT6; KMT6A; lysine N-methyltransferase 6; mKIAA4065; OTTHUMP00000213774; OTTHUMP00000213778; OTTHUMP00000213779; OTTHUMP00000213780; WVS; WVS2
Mouse
Protein G
RUO
2146
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.