missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EZH2/KMT6 Mouse anti-Human, Clone: EZH2/2536, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | EZH2/KMT6 |
|---|---|
| Clone | EZH2/2536 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Protein Array |
| Applications | ELISA, Peptide Array |
Description
EZH2/KMT6 Monoclonal specifically detects EZH2/KMT6 in Human samples. It is validated for Western Blot, Protein Array.Specifications
| EZH2/KMT6 | |
| 0.2 mg/mL | |
| ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, Chromatin Research, Phospho Specific, Prostate Cancer | |
| 10 mM PBS with 0.05% BSA | |
| 2146 | |
| IgG2a κ | |
| Protein A or G purified |
| EZH2/2536 | |
| Western Blot 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, Lysine N-methyltransferase 6, MGC9169 | |
| Recombinant full-length human EZH2/KMT6 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title