missing translation for 'onlineSavingsMsg'
Learn More

EZH2/KMT6 Antibody (EZH2/2536) - Azide and BSA Free, Novus Biologicals™

Product Code. 18339802 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339802 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339802 Supplier Novus Biologicals Supplier No. NBP308932

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EZH2/KMT6 Monoclonal specifically detects EZH2/KMT6 in Human samples. It is validated for Western Blot, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EZH2/KMT6
Applications Western Blot, Peptide Array
Classification Monoclonal
Clone EZH2/2536
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1-2 ug/mL, Protein Array
Formulation 10mM PBS
Gene Alias EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, Lysine N-methyltransferase 6, MGC9169
Host Species Mouse
Immunogen Recombinant full-length human EZH2/KMT6 protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, Phospho Specific, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2146
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.