missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EZH1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20852-1-AP-150UL
This item is not returnable.
View return policy
Description
EZH1 is a component of a noncanonical Polycomb repressive complex-2 (PRC2) that mediates methylation of histone H3 lys27 (H3K27) and functions in the maintece of embryonic stem cell pluripotency and plasticity (Shen et al., 2008 ).Specifications
| EZH1 | |
| Polyclonal | |
| Unconjugated | |
| EZH1 | |
| Enhancer of zeste homolog 1, ENX 2, EZH1, KIAA0388 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14055, 2145 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P70351, Q92800 | |
| EZH1 | |
| EZH1 Fusion Protein Ag14732 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction