missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EZH1 Polyclonal Antibody

Product Code. 12829282
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12829282 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12829282 Supplier Invitrogen™ Supplier No. PA521137

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is mouse lung tissue lysate. PA5-21137 can be used with blocking peptide PEP-1251.

EZH1 is a component of a noncanonical Polycomb repressive complex-2 (PRC2) that mediates methylation of histone H3 lys27 (H3K27) and functions in the maintenance of embryonic stem cell pluripotency and plasticity (Shen et al., 2008 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EZH1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene EZH1
Gene Accession No. P70351, Q92800
Gene Alias enhancer of zeste 1 polycomb repressive complex 2 subunit; enhancer of zeste homolog 1; enhancer of zeste homolog 1 (Drosophila); Enx2; ENX-2; Ezh1; Histone-lysine N-methyltransferase EZH1; hypothetical protein LOC664754; KIAA0388; KMT6B; si:ch211-250i3.1; zgc:136829
Gene Symbols EZH1
Host Species Rabbit
Immunogen An 18 amino acid peptide near the amino terminus of human EZH1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14055, 2145, 303547
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.