missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EZH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115205
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total EZH1.
EZH1 is a component of a noncanonical Polycomb repressive complex-2 (PRC2) that mediates methylation of histone H3 lys27 (H3K27) and functions in the maintenance of embryonic stem cell pluripotency and plasticity (Shen et al., 2008 ).
Specifications
| EZH1 | |
| Polyclonal | |
| Unconjugated | |
| EZH1 | |
| enhancer of zeste 1 polycomb repressive complex 2 subunit; enhancer of zeste homolog 1; enhancer of zeste homolog 1 (Drosophila); Enx2; ENX-2; Ezh1; Histone-lysine N-methyltransferase EZH1; hypothetical protein LOC664754; KIAA0388; KMT6B; si:ch211-250i3.1; zgc:136829 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14055, 2145 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P70351, Q92800 | |
| EZH1 | |
| A synthesized peptide derived from human EZH1(Accession Q92800), corresponding to amino acid residues S172-G222. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction