missing translation for 'onlineSavingsMsg'
Learn More

Ezetimibe Hydroxy tert-Butyldiphenylsilyl Ether, TRC

Product Code. 30395526
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395526 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395526 Supplier Toronto Research Chemicals Supplier No. TRCE97506050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ezetimibe Hydroxy tert-Butyldiphenylsilyl Ether
CAS 1217748-67-8
Molecular Formula C40H39F2NO3Si
Quantity 50 mg
Synonym (3R,4S)-1-(4-Fluorophenyl)-3-[(3S)-3-tert-butyldiphenylsilyloxy-3-(4-fluorophenyl)propyl]-4-(4-hydroxyphenyl)-2-azetidinone
InChI Formula InChI=1S/C40H39F2NO3Si/c1-40(2,3)47(34-10-6-4-7-11-34,35-12-8-5-9-13-35)46-37(28-14-18-30(41)19-15-28)27-26-36-38(29-16-24-33(44)25-17-29)43(39(36)45)32-22-20-31(42)21-23-32/h4-25,36-38,44H,26-27H2,1-3H3/t36-,37-,38-/m0/s1
SMILES CC(C)(C)[Si](O[C@@H](CC[C@H]1[C@@H](N(C1=O)c2ccc(F)cc2)c3ccc(O)cc3)c4ccc(F)cc4)(c5ccccc5)c6ccccc6
IUPAC Name (3S,4R)-3-[(3S)-3-[tert-butyl(diphenyl)silyl]oxy-3-(4-fluorophenyl)propyl]-1-(4-fluorophenyl)-4-(4-hydroxyphenyl)azetidin-2-one
Molecular Weight (g/mol) 647.82
Formula Weight 647.2667
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.