Learn More
Leica Microsystems Binocular EZTube™ Microscope Tube
Product Code. 12857905
Click to view available options
Includes:
Eyeguards
Product Code. 12857905
Brand: Leica Microsystems 13613323
This item requires a price update prior to being ordered.
Request a Price for this item here.
Outfit your DM300 Microscope with the viewing tube of your choice. Leica Microsystems™ Binocular EZTube™ Microscope Tube is precision crafted and constructed with quality Leica™ components.
  • For DM300 Microscope.
  • Includes eyeguard.
  • Available with or without pointer.
Specifications
Type EZTube™ Microscope Tube
For Use With (Equipment) DM300 Microscopes
Includes Eyeguards