missing translation for 'onlineSavingsMsg'
Learn More

(EZ)-N-[2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphinyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine, Mikromol™

Product Code. 30052238
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30052238

Brand: Mikromol MM0220.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(EZ)-N-[2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphinyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (EZ)-N-[2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphinyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine
CAS 102273-13-2
Analyte or Component Names (EZ)-N-[2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphinyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine
Shipping Condition Ice pack (-20°C)
Molecular Formula C12 H21 N5 O3 S2
Certifications/Compliance ISO 17025
Synonym (EZ)-N-[2-[[[2-[(Dimethylamino)methyl]thiazol-4-yl]methyl]sulphinyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine, Nizatidine Imp. C (EP), Nizatidine Impurity C
InChI Formula InChI=1S/C12H21N5O3S2/c1-13-11(6-17(18)19)14-4-5-22(20)9-10-8-21-12(15-10)7-16(2)3/h6,8,13-14H,4-5,7,9H2,1-3H3
SMILES CNC(=C[N+](=O)[O-])NCCS(=O)Cc1csc(CN(C)C)n1
IUPAC Name 1-N'-[2-[[2-[(dimethylamino)methyl]-1,3-thiazol-4-yl]methylsulfinyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine
Molecular Weight (g/mol) 347.46
Quantity 50 mg
Formula Weight 347.1086 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.