missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Hydrazide-Biotin

Product Code. 10027053 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10027053 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10027053 Supplier Thermo Scientific™ Supplier No. 21339

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biotinylate glycoproteins (aldehydes of oxidized sugars) with this simplest and shortest (15.7A) hydrazide-activated biotinylation reagent.

Thermo Scientific™ EZ-Link Hydrazide-Biotin is the shortest and simplest hydrazide-activated biotinylation reagent for labeling glycoproteins and other carbohydrate-containing compounds having oxidizable sugars or aldehydes.

Highlights:

  • Carbohydrate-reactive
  • Shorter spacer arm than Biotin-LC-Hydrazide
  • Must be dissolved in DMSO (50mM) before adding to aqueous buffer
  • Use at pH 4-6 to complete coupling reaction
  • Forms hydrazone bond
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hydrazide-Biotin
Quantity 100 mg
Content And Storage Store at 4°C.
Product Line EZ-Link
Cell Permeability Cell-Permeant
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Reactive Moiety Amine, Hydrazide
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Label or Dye Biotin
Spacer Short
Label Type Biotin & Analogs
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.