missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EYS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555507
This item is not returnable.
View return policy
Description
Immunogen sequence: CECTSGWTGQ NCSEEINECD SDPCMNGGLC HESTIPGQFV CLCPPLYTGQ FCHQRYNLCD LLHNPCR Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 46%.
Specifications
| EYS | |
| Polyclonal | |
| Unconjugated | |
| EYS | |
| bA166P24.2; bA307F22.3; bA74E24.1; C6orf178; C6orf179; C6orf180; dJ1018A4.2; dJ22I17.2; dJ303F19.1; EGFL10; EGFL11; EGF-like protein 10; EGF-like protein 11; EGF-like-domain, multiple 10; EGF-like-domain, multiple 11; Epidermal growth factor-like protein 10; epidermal growth factor-like protein 11; eyes shut homolog (Drosophila); EYS; Protein eyes shut homolog; protein spacemaker homolog; RP25; SPAM; UNQ9424/PRO34591 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 346007 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5T1H1 | |
| EYS | |
| Recombinant protein corresponding to Human EYS. Recombinant protein control fragment (Product #RP-94712). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction