missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Grommets extend the life of data tags by preventing tearing and abrasion.
- Black plastic grommets snap in easily to the pre-punched holes in thermal transfer printable tags from Brady.
- Allows tags to be customized or serially printed in a continuous manner on a thermal transfer printer before the grommet is applied
- Package includes grommets only (printer and tags sold separately).
Specifications
Specifications
| Product Type | Grommets |
| Color | Black |
| For Use With (Equipment) | BBP30 Label Printer, BBP31 Sign and Label Printer, BBP33 Label Printer, BBP35 Multi-Color Sign and Label Printer, BBP37 Color and Cut Sign and Label Printer, BBP85 Industrial Sign and Label Printer, i3300 Printer, S3000 Printer, S3100 Printer, GlobalMark Label Maker, PowerMark Industrial Sign and Label Printer |
| Description | Grommets (100-pack) |
| Quantity | 100 Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?