Learn More
EYA4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24691-1-AP-20UL
 View more versions of this product
Product Code. 16803185
  • £135.00 / 20µL
Description
This gene encodes a member of the eyes absent (EYA) family of proteins. The encoded protein may act as a transcriptional activator through its protein phosphatase activity, and it may be important for eye development, and for continued function of the mature organ of Corti. Mutations in this gene are associated with postlingual, progressive, autosomal domit hearing loss at the deafness, autosomal domit non-syndromic sensorineural 10 locus. The encoded protein is also a putative oncogene that mediates DNA repair, apoptosis, and innate immunity following DNA damage, cellular damage, and viral attack. Defects in this gene are also associated with dilated cardiomyopathy 1J. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
Specifications
EYA4
Polyclonal
Unconjugated
EYA4
CMD1J, DFNA10, EYA4, Eyes absent homolog 4
Rabbit
Antigen Affinity Chromatography
RUO
14051, 2070
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95677, Q9Z191
EYA4
EYA4 Fusion Protein Ag20466
20 μL
Primary
Mouse, Human