Learn More
EYA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562609
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: YGLPPFGALW PGMKPESGLI QTPSPSQHSV LTCTTGLTTS QPSPAHYSYP IQASSTNASL ISTSSTIANI PAAAVASISN QDYPTYTI Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
Eyes absent homolog 3 (EYA3) is a member of the eyes absent (EYA) family of proteins. This protein may act as a transcriptional activator and have a role during development. A similar protein in mice can act as a transcriptional activator. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
EYA3 | |
Polyclonal | |
Unconjugated | |
Eya3 | |
AI844637; EYA transcriptional coactivator and phosphatase 3; EYA3; eyes absent 3; eyes absent 3 homolog; eyes absent 3 homolog (Drosophila); eyes absent homolog 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2140 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99504 | |
Eya3 | |
Recombinant protein corresponding to Human EYA3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only