missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EYA3 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46819R
This item is not returnable.
View return policy
Description
EYA3 Polyclonal antibody specifically detects EYA3 in Human samples. It is validated for Western Blot
Specifications
| EYA3 | |
| Polyclonal | |
| Western Blot | |
| DKFZp686C132, EC 3.1.3.48, eyes absent (Drosophila) homolog 3, eyes absent 3, eyes absent homolog 3, eyes absent homolog 3 (Drosophila) | |
| The immunogen for this product maps to a region between residue 30 and 60 of human eyes absent homolog 3 using the numbering given in entry NP_001981.2 (GeneID 2140). | |
| 0.1 mL | |
| Vision | |
| 2140 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction