missing translation for 'onlineSavingsMsg'
Learn More

EYA3 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18402786 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402786 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402786 Supplier Novus Biologicals Supplier No. NBP146819AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EYA3 Polyclonal antibody specifically detects EYA3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen EYA3
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZp686C132, EC 3.1.3.48, eyes absent (Drosophila) homolog 3, eyes absent 3, eyes absent homolog 3, eyes absent homolog 3 (Drosophila)
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 30 and 60 of human eyes absent homolog 3 using the numbering given in entry NP_001981.2 (GeneID 2140).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 2140
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.