missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ EYA2 Recombinant Rabbit Monoclonal Antibody (K01_2M33)
Beskrivelse
EYA2 Recombinant Monoclonal Antibody for Western Blot
Functions both as protein phosphatase and as transcriptional coactivator for SIX1, and probably also for SIX2, SIX4 and SIX5. Tyrosine phosphatase that dephosphorylates 'Tyr- 142' of histone H2AX (H2AXY142ph) and promotes efficient DNA repair via the recruitment of DNA repair complexes containing MDC1. 'Tyr-142' phosphorylation of histone H2AX plays a central role in DNA repair and acts as a mark that distinguishes between apoptotic and repair responses to genotoxic stress. Its function as histone phosphatase may contribute to its function in transcription regulation during organogenesis. Plays an important role in hypaxial muscle development together with SIX1 and DACH2; in this it is functionally redundant with EYA1.
Tekniske data
Tekniske data
| Antigen | EYA2 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2M33 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | EYA2 |
| Gene Accession No. | O00167 |
| Gene Alias | Drosophila-type eyes absent 2-like protein; Eab1; EYA transcriptional coactivator and phosphatase 2; Eya2; eyes absent 2; eyes absent 2 homolog; eyes absent 2 homolog (Drosophila); eyes absent 2 isoform a; eyes absent homolog 2 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?