missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EYA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102947
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total EYA2.
Functions both as protein phosphatase and as transcriptional coactivator for SIX1, and probably also for SIX2, SIX4 and SIX5. Tyrosine phosphatase that dephosphorylates 'Tyr- 142' of histone H2AX (H2AXY142ph) and promotes efficient DNA repair via the recruitment of DNA repair complexes containing MDC1. 'Tyr-142' phosphorylation of histone H2AX plays a central role in DNA repair and acts as a mark that distinguishes between apoptotic and repair responses to genotoxic stress. Its function as histone phosphatase may contribute to its function in transcription regulation during organogenesis. Plays an important role in hypaxial muscle development together with SIX1 and DACH2; in this it is functionally redundant with EYA1.
Specifications
| EYA2 | |
| Polyclonal | |
| Unconjugated | |
| EYA2 | |
| Drosophila-type eyes absent 2-like protein; Eab1; EYA transcriptional coactivator and phosphatase 2; Eya2; eyes absent 2; eyes absent 2 homolog; eyes absent 2 homolog (Drosophila); eyes absent 2 isoform a; eyes absent homolog 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 156826, 2139 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00167 | |
| EYA2 | |
| A synthesized peptide derived from human EYA2(Accession O00167), corresponding to amino acid residues V203-S253. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction