missing translation for 'onlineSavingsMsg'
Learn More

EYA2 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18401857 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401857 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401857 Supplier Novus Biologicals Supplier No. NBP146817AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EYA2 Polyclonal antibody specifically detects EYA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen EYA2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EAB1EC 3.1.3.48, eyes absent (Drosophila) homolog 2, eyes absent homolog 2, eyes absent homolog 2 (Drosophila), MGC10614
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 100 and 150 of human eyes absent homolog 2 using the numbering given in entry NP_005235.3 (GeneID 2139).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2139
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.