missing translation for 'onlineSavingsMsg'
Learn More

Manutan Extra tubular shelf for tubular shelving, 1250mm x 400mm

Product Code. 12859128
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12859128 Supplier Manutan Supplier No. A360731
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Strong and durable additional tubular shelf, equipped with welded hooks for easy connection to uprights. Ideal for storing larger items. Easy click-on system with welded hooks for easy connection to uprights.

  • Grey
  • Shelf capacity UDL: 115kg
  • Dimensions (W x D): 1250mm x 400mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.