missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Extra tubular shelf for tubular shelving, 1000mm x 3700mm
Description
Strong and durable additional tubular shelf, equipped with welded hooks for easy connection to uprights. Ideal for storing larger items. Easy click-on system with welded hooks for easy connection to uprights.
- Grey
- Shelf capacity UDL: 233kg
- Dimensions (W x D): 1000mm x 700mm
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?