missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXTL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112818
This item is not returnable.
View return policy
Description
EXTL3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
EXTL3 is probable glycosyltransferase.
Specifications
| EXTL3 | |
| Polyclonal | |
| Unconjugated | |
| EXTL3 | |
| 2900009G18Rik; AU040105; BOTV; exostoses (multiple)-like 3; exostoses-like 3; exostosin; exostosin like glycosyltransferase 3; exostosin tumor-like 3; exostosin-like 3; exostosin-like glycosyltransferase 3; Ext1l; EXTL1L; EXTL3; EXTR1; EXT-related 1; EXT-related protein 1; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; hereditary multiple exostoses gene isolog; KIAA0519; mKIAA0519; multiple exostosis-like protein 3; Putative tumor suppressor protein EXTL3; REG; reg receptor; REGR; RPR | |
| Rabbit | |
| Protein G | |
| RUO | |
| 2137 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O43909 | |
| EXTL3 | |
| Recombinant Human Exostosin-like 3 protein (2-245AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido