missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXTL2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148601
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
EXTL2 is a glycosyltransferase required for the biosynthesis of heparan-sulfate and responsible for the alternating addition of beta-1-4-linked glucuronic acid (GlcA) and alpha-1-4-linked N-acetylglucosamine (GlcNAc) units to nascent heparan sulfate chains.
Specifications
| EXTL2 | |
| Polyclonal | |
| Unconjugated | |
| EXTL2 | |
| 3000001D04Rik; alpha-1,4-N-acetylhexosaminyltransferase EXTL2; Alpha-GalNAcT EXTL2; AW146439; exostoses (multiple)-like 2; exostoses-like 2; exostosin like glycosyltransferase 2; exostosin-like 2; exostosin-like glycosyltransferase 2; exotoses (multiple)-like 2; EXTL2; Extr2; EXT-related protein 2; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; processed exostosin-like 2 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 58193 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9ES89 | |
| EXTL2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Exostosin-like 2/EXTL2 Lumenal Domain, Asn43-Met330. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction