Learn More
Grant Instruments™ External Probe
TX external probes (optional) for monitoring and controlling temperature of remote loads.
Brand: Grant Instruments™ TXSEP
Code : 03
Additional Details : Weight : 0.50000kg
Description
- Flexible plastic or Stainless steel probe
- 3m cable (Din plug)
Specifications
3m cable (DIN plug) | |
Stainless Steel |