missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Extensor Long PCR Enzyme Mix Products

Product Code. 11819512
Change view
Click to view available options
Description:
Extensor Master Mix (Buffer 1 Formulation)
Extensor ReddyMix Master Mix (Buffer 1 Formulation)
Quantity:
400 x 25µL Rxns
500 25µL Rxns
Unit Size:
400 reactions
Product Code. Description Quantity unitSize
11819512 Extensor Master Mix (Buffer 1 Formulation) 500 25µL Rxns 400 reactions
12340130 Extensor ReddyMix Master Mix (Buffer 1 Formulation) 400 x 25µL Rxns 400 reactions
2 options
This item is not returnable. View return policy

Product Code. 11819512

Brand: Thermo Scientific™ AB0792/B

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Amplify long PCR products up to 20kb with this synergistically-acting PCR enzyme blend of Taq DNA polymerase and a proprietary proofreading enzyme.

Extensor Long Range PCR Enzyme is a blend of ThermoPrime Taq DNA Polymerase and a proprietary thermostable proofreading enzyme. The two enzymes act synergistically to generate long PCR products, up to 20 kb. Extensor Long Range Enzyme includes 2 buffer formulations; Buffer 1 for templates up to 12 kb in length, and Buffer 2 for templates 12 to 20 kb and GC-rich or other difficult templates. This enzyme blend is also available in 2X Master Mix and ReddyMix formulations. The ReddyMix Master Mix contains an inert red dye and a density reagent that allows for direct loading onto agarose gels.

Extensor Long Range Enzyme Blend is an optimized mix of ThermoPrime™ DNA Polymerase and a proprietary thermostable proofreading enzyme. The two enzymes act synergistically to generate long PCR products, up to 20kb. This makes the enzyme blend ideal for robust full-length amplification of long range sequences.
  • Amplifies targets up to 20kb in length
  • Unique enzyme blend and optimized buffer combination delivers increased accuracy and yield
  • PCR products generated can be used directly in T/A cloning
  • Optimized buffering systems for success with a variety of templates
  • Buffer 1 for success with most templates up to 12kb in length
  • Buffer 2 for templates 12-20kb and GC-rich or other difficult templates of any length
Extensor is available in a range of formats and pack sizes to suit any workflow.

Format Options:

  • Enzyme Kits are the most basic format. Kits include the enzyme, PCR buffers 1 and 2, and 25mM MgCl2 solution, each in separate vials. dNTPs must be purchased separately

  • Complete PCR Kits include all the components contained in the Enzyme Kit, plus they have HPLC-purified dNTP mix. Complete PCR kits are a convenient option that provides all the components necessary for PCR

  • Master Mix formats consist of a single master buffer containing all the components needed for PCR. Master Mixes are the most convenient format and significantly reduce assay set-up steps

  • ReddyMix Master Mix formats consist of a single ReddyMix master buffer containing all the components needed for PCR. ReddyMix master mix can be directly loaded onto agarose gels post-PCR

  • PCR Buffers: All kits are supplied with both Buffer 1 and Buffer 2 to accommodate a variety of template types. Master mixes come in either Buffer 1 or Buffer 2 formulation

Recommended for:

Amplifying fragments up to 20kb; PCR; Cloning; Long PCR

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Store at -20°C. Avoid freeze/thaw cycles
Description Extensor Master Mix (Buffer 1 Formulation)
Quantity 500 25µL Rxns
For Use With (Application) Cloning

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.