Learn More
Memmert™ Extended Overtemperature Protection for UF160
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF160 Oven
Unit Size:
Each
Description
- Accessory
- With integrated Pt100 sensor
- For models with SingleDISPLAY
Specifications
Specifications
| For Use With (Equipment) | UF160 Oven |
| Includes | Integrated Pt100 Sensor |