Learn More
Thermo Scientific™ Extended Length 200μL ART™ Barrier Specialty Filter Pipettor Tips
Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
£205.00
Specifications
Description | ART XLP 200, Filtered, Sterile, |
---|---|
Volume (Metric) | 200 μL |
Filtered | Filtered |
Sterility | Sterile |
Color | Natural |
Description
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.
Features:
- Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
- Available in sterile, nonsterile and in various packaging options
- Manufactured according to the industry's highest quality standard
Designed to aid in pipetting for a variety of applications:
Extended length to prevent contamination by keeping the pipette shaft from entering the sample vessel
Gel loading for loading acrylamide or agarose gels
Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Solvent Safe for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson Pipetman™ ™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
Specifications
ART XLP 200, Filtered, Sterile, | |
Filtered | |
Natural | |
3.59 in. | |
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free | |
Extended Length | |
ART |
200 μL | |
Sterile | |
ART XLP 200 | |
9.12cm | |
Rack | |
Universal Fit | |
Sterile & PCR Ready |