missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EXPRESS qPCR Supermix, universal

Product Code. p-7083357
Click to view available options
No. of Reactions:
200 Reactions
2500 Reactions
Unit Size:
1 set
2500 reactions
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10657133

Brand: Invitrogen™ 11785200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for probe-based qPCR assays.

  • EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for probe-based qPCR assays
  • This kit comes with a separate tube of ROX reference dye
  • EXPRESS qPCR SuperMix kits with premixed ROX reference dye are also available

This kit provides

  • Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
  • UDG carryover protection—uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination

Order Info

Guaranteed minimum shelf life is 60 days.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage EXPRESS qPCR SuperMix kits are provided as a 2X qPCR SuperMix with ROX premixed or in a separate tube.

Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteedstable for 6 months when properly stored.
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7500 Fast System, 7500 System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, BioRad CFX96™, Eppendorf™ MasterCycler™ RealPlex, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3005P, Stratagene Mx4000, BioRad MyiQ, Corbett RotorGene, Cepheid SmartCycler, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, BioRad iCycler iQ
Product Type Express qPCR Supermix
Quantity 200 Reactions
Sufficient For 200 Reactions
For Use With (Application) Gene Expression
No. of Reactions 200 Reactions
Sample Type DNA (Genomic), cDNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.