missing translation for 'onlineSavingsMsg'
Learn More

SuperScript™ EXPRESS One-Step Superscript™ qRT-PCR Kit, universal

Product Code. 10449213
Change view
Click to view available options
No. of Reactions:
2500 Reactions
500 Reactions
Unit Size:
2500 reactions
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
10449213 500 Reactions Each
10782134 2500 Reactions 2500 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10449213 Supplier SuperScript™ Supplier No. 11781200
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Kit incorporates proven Invitrogen™ enzymes specifically tailored to deliver superior results on high-throughput, fast-cycling instruments for probe-based qPCR experiments.

  • Kit incorporates proven Invitrogen™ enzymes specifically tailored to deliver superior results on high-throughput, fast-cycling instruments for probe-based qPCR experiments
  • This fast, one-step mix is designed to work with a variety of instruments’ default protocols, including Applied Biosystems and non-Applied Biosystem (see universal version) qPCR platforms
  • Fast-activating, antibody-mediated Platinum Taq DNA Polymerase for complete, rapid activation; provides high sensitivity and reproducibility
  • The One-Step SuperScript III Reverse Transcriptase module is a unique formulation to ensure high yield and less PCR inhibition
  • Flexible formats with separate or pre-mixed ROX are designed to work with default instrument protocols on a wide range of high-throughput fast instruments
  • Contains heat-labile UDG which significantly reduces carryover contamination risk with excellent activity from room temperature to approximately 40°C
  • Due to the thermostability of SuperScript III, the cDNA synthesis step is run at 50°C, at which temperature the heat-labile UDG is totally and irreversibly inactivated
  • Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at 4°C to 8°C
  • Store at -20°C for long-term storage
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The EXPRESS One-Step SuperScript™ qRT-PCR Universal kit contains 3 modules: 2X EXPRESS SuperScript™ Mix (1 ml), Express qPCR SuperMix Universal (5 ml), and ROX Reference Dye (500 μL).

Store at -20°C for long-term storage. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at 4–8°C.
Detection Method Primer-probe
Format Tube
PCR Method 1-step RT-qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7500 Fast System, 7500 System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, BioRad CFX96™, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, BioRad MyiQ, Corbett RotorGene, Cepheid SmartCycler, MJ Opticon, MJ Chromo4, BioRad iCycler iQ
Product Line SuperScript
Product Type qRT-PCR Kit
Quantity 500 reactions
Sufficient For 500 Reactions
No. of Reactions 500 Reactions
Sample Type RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.