missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Exportin-T Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29009
This item is not returnable.
View return policy
Description
Exportin-T Polyclonal antibody specifically detects Exportin-T in Human,Mouse samples. It is validated for Western Blot
Specifications
| Exportin-T | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| exportin(tRNA), exportin, tRNA (nuclear export receptor for tRNAs), exportin-T, tRNA exportin, XPO3 | |
| Between 825 and 875 | |
| 100 μg | |
| Signal Transduction | |
| 11260 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction