missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exportin-5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | Exportin-5 |
|---|---|
| Dilution | Western Blot 1:2000 - 1:10000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231196
|
Novus Biologicals
NBP3-38333-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231946
|
Novus Biologicals
NBP3-38333-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Exportin-5 Polyclonal antibody specifically detects Exportin-5 in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| Exportin-5 | |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Epigenetics | |
| PBS (pH 7.3), 50% glycerol | |
| 57510 | |
| IgG | |
| Affinity purified |
| Western Blot 1:2000 - 1:10000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Exp5, exportin 5, exportin-5, FLJ14239, FLJ32057, FLJ45606, KIAA1291exp5, Ran-binding protein 21, RANBP21 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 925-1204 of human Exportin-5 (Exportin-5 (XPO5)) (NP_065801.1).,, Sequence:, MRLSQKWQVINQRSLLCGEDEAADENPESQEMLEEQLVRMLTREVMDLITVCCVSKKGADHSSAPPADGDDEEMMATEVTPSAMAELTDLGKCLMKHEDVCTALLITAFNSLAWKDTLSCQRTTSQLCWPLLKQVLSGTLLADAVTWLFTSVLKGLQMHGQHDGCMASLVHLAFQIYEALRPRYLEIRAVMEQIPEIQKDSLDQFDCKLLNPSLQKVADKRRKDQFKRLIAGCIGKPLGEQFRKEVHIKNLPSLFKKTKPMLETEVLDNDGGGLATIFEP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title