missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ ExpiFectamine™ 293 Met (–) Transfection Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
Kit components are specifically formulated without methionine, allowing users to add labeled methionine of their choice.
Featues:
- Methionine-free formulation
- 2- to 10-fold higher protein yields compared to other transfection reagents used for high-density 293 cell culture
- Same transient transfection protocol as ExpiFectamine 293 Transfection Kit
- Scalable transfection from small to large scale while maintaining equivalent volumetric protein yields
Specifically designed to work with Expi293F cell lines as part of the Expi293 Expression System
Components included in the ExpiFectamine 293 Met (–) Transfection Kit are:
- 2.7 mL ExpiFectamine 293 Reagent
- 5 mL ExpiFectamine 293 Transfection Enhancer
- 1 x 50 mL ExpiFectamine 293 Met (–) Transfection Enhancer 2
- 100 mL Opti-Plex Complexation Buffer (methionine-free)
Component volumes are sufficient to transfect 1 L of culture
Specifications
Specifications
| Classification | AOF, Chemically Defined, Methionine-Free, Protein-Free, Serum Free |
| Content And Storage | • 3 x 0.9 mL ExpiFectamine 293 Transfection Reagent, store at 2–8°C • 5 mL ExpiFectamine 293 Enhancer 1, store at 2–8°C • 50 mL ExpiFectamine 293 Met (–) Enhancer 2, store at 2–8°C • 100 mL Opti-Plex Complexation Buffer, store at 2–8°C (protect from light) |
| Cell Type | Established Cell Lines |
| For Use With (Application) | Transfection |
| Product Line | ExpiFectamine™ |
| Product Type | Transfection Kit |
| Quantity | 1 Kit |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet or Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction