missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ ExpiCHO™ Stable Production AGT Medium

Product Code. 30282638
Change view
Click to view available options
Quantity:
10 L
100 L
450 L
Unit Size:
10 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30282638 450 L -
30284703 10 L 10 L
30283516 100 L -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30282638 Supplier Gibco™ Supplier No. A3711103

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco ExpiCHO Stable Production Medium (SPM) is specifically designed to simplify your cell line development process into production with minimal effort and a high degree of confidence.

Gibco ExpiCHO Stable Production Medium (SPM) is specifically designed to simplify your cell line development process into production with minimal effort and a high degree of confidence. This chemically defined, protein-free, animal origin component-free formulation is fully optimized for large-scale fed-batch culture with ExpiCHO-S cells. ExpiCHO SPM supports an easy transition from transient to stable production, provides high titers without the need for additional medium optimization (see figures below), and works well with existing EfficientFeed C+ supplement.

ExpiCHO SPM is part of the ExpiCHO family, an end-to-end solution that empowers you to carry out your entire workflow from transient to stable expression without having to screen or optimize media and feeds.

Hassle-free scale-up

Simplify your process as you transition into stable clones with ExpiCHO SPM—no cell adaptation, no media optimization, no changes in cell doubling time and peak density. ExpiCHO SPM eases the transition using ExpiCHO-S cells and provides optimal performance with less need for fine-tuning from development scientists to get you to the next step faster. This medium supports high titers under standard fed-batch process conditions in conjunction with EfficientFeed C+ supplement. It is available in liquid format (A3711001) or in our AGT dry format (this page) from 10 L through 450 L equivalents with larger sizes available upon request.

ONE Solution

The ExpiCHO family includes products that are engineered to work seamlessly with each other. ExpiCHO SPM is a member of this family and allows expanded support of bioproduction from transient to stable protein expression. Look for ExpiCHO-branded protocols and methods to get optimal performance in the development process.

Members of the ExpiCHO family include:

  • ExpiCHO Stable Production Medium (this page)
  • ExpiCHO Expression Medium (A2910001)
  • ExpiCHO-S cells (A29127)
  • Expifectamine CHO Transfection Kit (A29130)

The power to streamline transfer to manufacturing

ExpiCHO SPM liquid format and AGT dry format demonstrate comparable cell growth and titer performance (see figure below). Therefore, scale-up and manufacturing using AGT dry format is simplified by knowing that the reconstituted AGT dry format will behave similarly to the liquid format used prior to scale-up. The AGT dry format is easy to reconstitute; just add water and it will automatically adjust pH and osmolality. This can help provide efficiencies in supply chain complexity, reconstitution complexity, and, therefore, overall expense.

When to use ExpiCHO Stable Production Medium:

  • When you are using the ExpiCHO-S cell line for transient expression and want to scale up
  • After you have completed the stable transfection of ExpiCHO-S cells in ExpiCHO Expression Medium. (Follow our recommended protocol.)
  • When you do not have the time to perform medium adaptation or optimization

When NOT to use ExpiCHO Stable Production Medium:

  • When you have an existing platform cell-line or feeds and are looking for a broad medium to support a wide variety of cell-lines
  • When you need a medium for transfection. For transient and stable transfection, ExpiCHO Expression Medium is recommended.

Support available with ExpiCHO Stable Production Medium:

  • A stable cloning protocol designed specifically for the ExpiCHO family of products (must be used for optimal performance)
  • Licensing for the ExpiCHO-S cells for development of stable clones. (Contact us at outlicensing@thermofisher.com.)

Design your own media specific to your needs

Whether it's a bench-level experiment or large-scale bio-manufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco BioProduction Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you. Reach out for consultation and/or services in medium, feed, or cell line development.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line ExpiCHO-S
Classification Animal Origin-free, Chemically-defined, Protein-free
Form Powder (AGT)
Product Type ExpiCHO Stable Production Medium (SPM)
Product Line ExpiCHO
Quantity 450 L
Shipping Condition Approved for shipment on Wet or Dry Ice
Format Ropack Bucket
Content And Storage Store in refrigerator (2–8°C).
When subculturing ExpiCHO the day before a transfection to 3-4 x 10E6/mL, would you recommend pelleting the cells and resuspending them in completely fresh media, or simply replacing some of the media to split them down to the desired cell density?

Before transfecting ExpiCHO cells, there is no need to change out the media during the subculturing. This would become expensive and the conditioned media is not necessary.

For how many passages would you recommend using ExpiCHO cells before replacing them with new cells that were frozen at earlier passages?

For optimal performance with respect to protein production, we recommend thawing ExpiCHO cells and letting them grow for 2-3 passages prior to transfection. Overall, we do not recommend using the cells beyond ˜20 passages.

Is the ExpiCHO Stable Production Medium (SPM) compatible with the GS/MSX (Glutamine Synthetase Expression Technology) selection?

Yes, ExpiCHO Stable Production Medium (SPM) is compatible with the GS/MSX (Glutamine Synthetase Expression Technology) selection.

How much time do I need to consider for the stability study using ExpiCHO Stable Production Medium (SPM)?

After having obtained best expressing clones, a stability study is done. The time for a stability study varies (it depends how fast the cells double). It takes 60 generations or two months (whichever comes first).

When performing stable protein production using ExpiCHO Stable Production Medium (SPM), how is a stability study performed?

A stability study involves testing of ExpiCHO-S clones for 60 generations (60 population doublings) to see if high protein expression is maintained.

Is the transfection step different when stable selection is desired and ExpiCHO Stable Production Medium (SPM) is part of the workflow?

Yes, when stable selection is desired, transfection takes place in the presence of ExpiCHO Expression Medium but in the absence of the enhancer and feed. About 48 hours post-transfection, stable selection starts by addition of the appropriate selection antibiotics.

When should I not use ExpiCHO Stable Production Medium (SPM)?

We don't recommend using ExpiCHO SPM when you have an existing platform cell-line or feeds and are looking for a broadly usable medium, or when you need a transfection medium. We recommend using ExpiCHO Expression Medium for transfection.

When should I use ExpiCHO Stable Production Medium (SPM)?

We recommend using ExpiCHO SPM when transfection of ExpiCHO-S cells has been already completed with ExpiCHO Expression Medium, and you want to develop stable clones and scale up protein production.

What vector do you recommend using with ExpiCHO Stable Production Medium (SPM)?

We recommend using our pcDNA3.3-TOPO TA and pcDNA3.4-TOPO TA vectors. These vectors have only a single gene integration site so some modification will be necessary for antibody applications. You can use our GeneArt Express Cloning Service (https://www.thermofisher.com/us/en/home/life-science/cloning/gene-synthesis/geneart-plasmid-services/geneart-subcloning-service.html#express) to create a modification that will allow you to clone both the heavy and light chains into the vector (ideally joined by an IRES site). Please note that other vectors may also be used, and that you may want to test initially with 2-3 vectors for best results.

What vector did you use to produce the protein expression data with ExpiCHO Stable Production Medium (SPM)?

In our initial work, we used a proprietary vector that cannot be disclosed.

What is the difference between ExpiCHO Expression Medium and ExpiCHO Stable Production Medium (SPM)?

Both products are similar, however, formulation changes had to be made to make the ExpiCHO Stable Production Medium (SPM) scalable to AGT format. One such change is that it requires the addition of Glutamax whereas ExpiCHO Expression Medium does not. Additionally, ExpiCHO SPM is not compatible with transfection whereas ExpiCHO Expression Medium is compatible with transfection.

What is the intended use statement for ExpiCHO Stable Production Medium (SPM)?

The intended use statement for ExpiCHO Stable Production Medium (SPM) is as foll:
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Does ExpiCHO Stable Production Medium (SPM) contain Glutamax or Glutamine?

No. ExpiCHO Stable Production Medium (SPM) needs to be supplemented with Glutamax or Glutamine.

Can I perform transient transfection with ExpiCHO Stable Production Medium (SPM)?

No. Unlike ExpiCHO Expression Medium, ExpiCHO Stable Production Medium (SPM) is not recommended for transfection as it does not support high transient transfection efficiency.

What is ExpiCHO Stable Production Medium (SPM)?

ExpiCHO Stable Production Medium (SPM) is a chemically defined protein-free, animal origin component-free medium developed specifically to support high titer expression of stable ExpiCHO-S clones in suspension. The medium is designed to provide a seamless scale-up solution for customers using the ExpiCHO Expression System for transient production looking to transition to the development of stable ExpiCHO clones. The medium is formulated without hypoxanthine and thymidine for use in dihydrofolate reductase (DHFR)-amplified systems, without L-glutamine or GlutaMAX I Supplement for use in glutamine synthetase systems, and without phenol red in order to minimize the estrogen-like effects of phenol red.

ExpiCHO SPM allows for direct transition from transient expression to the large scale production of stable clones with no adaptation. You can start process development faster, and streamline or simplify transfer to manufacturing scale. ExpiCHO SPM is available in liquid format (Cat. No. A3711001) or in dry AGT format (Cat. Nos. A3711101, A3711102, and A3711103)

Note: ExpiCHO SPM is not compatible for use as a medium during the transfection step.

Can ExpiCHO-S Cells be used to create stable clones?

Our ExpiCHO-S Cells are derived from our cGMP banked CHO-S cells (Cat. No. A1155701), thus stable CHO-S selection methods are applicable to these cells. We recommend you follow the protocol outlined in the User Bulletin for the Creation and Scale up of a stable cell ine using ExpiCHO Products (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017764_CreatScaleup_StableCellExpiCHO_UB.pdf)


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.