missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ExpiCHO-S™ Cells (cGMP Banked)
ExpiCHO-S Cells are derived from a non-engineered sub-clone that was screened and isolated from CHO-S Chinese hamster ovary (CHO) cells. They are a core component of the ExpiCHO Expression System Kit.
Brand: Gibco A37785
Additional Details : Weight : 2.00000kg
Description
- ExpiCHO-S Cells are derived from a non-engineered sub-clone that was screened and isolated from CHO-S Chinese hamster ovary (CHO) cells
- They are a core component of the ExpiCHO Expression System Kit
- ExpiCHO-S Cells are maintained in suspension culture, have minimal tendency to clump, and grow to high density in ExpiCHO Expression Medium
- As part of the ExpiCHO Expression System Kit, the cells generate superior protein yields by transient transfection compared to standard CHO, 293, and high-density Expi293F cells
- Cells are derived from the same Master Seed Bank as standard ExpiCHO-S Cells
- Frozen cells are supplied in a vial containing approximately 1.1 mL of cells at 1 x 107 viable cells per mL in ExpiCHO Expression Medium and 10% DMSO
- The cells should be thawed directly into ExpiCHO Expression Medium
Specifications
Store in liquid nitrogen. | |
Suspension Cell Culture | |
Hamster | |
ExpiCHO-S™ |
ExpiCHO-S, cGMP Banked | |
Frozen | |
1 x 107 cells/mL | |
ExpiCHO-S Cells |