missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Expi293™ PRO Expression System Kit

Product Code. 31337525
Change view
Click to view available options
Quantity:
1 Starter Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31337525 1 Starter Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31337525 Supplier Gibco™ Supplier No. A40003643
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The Expi293 PRO Expression System enables fast, high-yield protein production from HEK293 cells. Designed for efficiency and scalability, it delivers superior expression of complex proteins with a simplified, automation-friendly workflow.

The Gibco Expi293 PRO Expression System is a complete solution for rapid, high-yield protein production from HEK293 cells. It is a fully integrated expression system designed to deliver higher yields of a wider variety of proteins—including challenging and low-yield proteins—with exceptional speed, efficiency, and an easy-to-automate protocol. Producing increasingly complex proteins with higher yields and increased throughput is critical to accelerating breakthroughs in protein research applications and drug development.

The Expi293 PRO Expression System features include:

  • Unlock new protein targets; express challenging and low-yield proteins
  • Exceptional protein yields; multi-fold increase in protein yields—up to 5 g/L
  • Cost effective; the lowest cost/mg
  • Rapid production of recombinant proteins in as little as 2–4 days
  • High throughput and automation compatible
  • High density cultures of Expi293 PRO cells that result in more expressing cells per milliliter of culture
  • Reduce reagents, plasticware, and labor by 3X
  • Native folding and mammalian post-translational modification of expressed proteins
  • Scalable expression from <1 mL to >10 L

The components included in the Expi293 PRO Expression System Kit are: two vials of frozen Expi293 PRO cells, one liter of Expi293 PRO Expression Medium, one Expi293 PRO Transfection Kit sufficient to transfect 1 liter of culture, Opti-Plex Complexation Buffer, and an antibody-expressing positive control vector. All components of the Expi293 PRO Expression System are also available for individual purchase.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Expi293 PRO
Cell Type HEK293
Format Kit
Promoter pcDNA™ 3.4
Transfection Technique Lipid-based Transfection, Transient Transfection
Species Human
Product Type Expression System Kit
Quantity 1 Starter Kit
Sterility Sterile

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.