missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Expi293™ PRO Cells

Product Code. 31337526 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 mL
6 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31337526 1 mL
31337533 6 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31337526 Supplier Gibco™ Supplier No. A40001140
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Expi293 PRO Cells are a next-generation HEK293 cell line for fast, high-yield protein expression. They support robust, scalable performance across a broad range of proteins and formats with improved growth kinetics and expression consistency up to 40 passages.

Expi293 PRO Cells are derived from the HEK 293 cell line and are a core component of the Expi293 PRO Expression System. They are maintained in suspension culture and grow to high density in Expi293 PRO Expression Medium. Expi293 PRO Cells are highly transfectable and generate superior protein yields compared to standard HEK 293 cell lines in transient protein expression.

Attributes of Expi293 PRO Cells include:

  • Suspension culture cells pre-adapted to Expi293 PRO Expression Medium
  • Higher protein expression levels than other HEK 293 cells adapted to Expi293 PRO Expression Medium
  • More rapid cell growth and higher culture viabilities than standard suspension-adapted HEK 293 cells
  • Consistent transient expression performance for 40 passages beyond thaw

Frozen cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in Expi293 PRO Expression Medium and 10% DMSO. The cells should be thawed directly into Expi293 PRO Expression Medium. The six-vial multipack eliminates the need to expand and rebank the cells in the lab.

The cells are typically grown in flasks on a shaker platform in a humidified, CO2 cell culture incubator. However, cultures are scalable from volumes of <1 mL in multiwell plates to >10 L in bioreactors. Protein harvest can be performed at 2–7 days post-transfection, depending on the protein expressed. For expression of recombinant antibodies, harvest is typically at 4–7 days post-transfection. There is no need for culture medium supplementation or batch feeding during expression.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 107 cells⁄mL, store in liquid nitrogen
Age Embryonic
Cell Line 293 (HEK)
Cell Type Kidney (Embryonic)
Culture Type Suspension Cell Culture
Form Cryopreserved
Gender Female
Species Human
Tissue Human Embryonic Kidney Tissue
Format Vial
Product Line Expi293 PRO
Product Type HEK293 Cells
Quantity 1 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.