missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ EXP 3174

Product Code. 18731799
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18731799

Brand: Tocris Bioscience™ 5728/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective AT1 receptor antagonist

EXP 3174 is a potent and selective noncompetitive AT1 receptor antagonist (IC50 = 37 nM). Reduces blood pressure in a rat model. Active metabolite of Losartan potassium (Cat. No. 3798).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Butyl-4-chloro-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic acid
CAS 124750-92-1
Quantity 10 mg
Target Angiotensin AT1 Receptor Antagonists
Molecular Formula C22H21ClN6O2
Formula Weight 436.89
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.