missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exostosin 1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £366.00
Specifications
| Antigen | Exostosin 1 |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18308646
|
Novus Biologicals
NBP3-03736-20UL |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18301754
|
Novus Biologicals
NBP3-03736-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Exostosin 1 Polyclonal specifically detects Exostosin 1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Exostosin 1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 2.4.1.224, EC 2.4.1.225, exostoses (multiple) 1, exostosin 1, exostosin-1, EXT, Glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase, Glucuronosyl-N-acetylglucosaminyl-proteoglycan/N-acetylglucosaminyl-proteoglycan4-alpha-N-acetylglucosaminyltransferase, Langer-Giedion syndrome chromosome region, LGCR, LGS, Multiple exostoses protein 1, N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase, Putative tumor suppressor protein EXT1, TRPS2, TTV | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 467-746 of human Exostosin 1 (NP_000118.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS with 50% glycerol, pH7.3. | |
| 2131 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title