missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Exosome Standards (PC3 cell line)

Product Code. 18614320 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 x 100 μg Vials
2 x 30 μg Vials
6 x 100 μg Vials
6 x 30 μg Vials
Unit Size:
Pack of 2
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18614320 2 x 30 μg Vials Pack of 2
18612660 6 x 100 μg Vials Pack of 6
18624320 2 x 100 μg Vials Pack of 2
18602041 6 x 30 μg Vials Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18614320 Supplier Novus Biologicals™ Supplier No. NBP24985660ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C. After reconstitution store at -70°C.
Concentration LYOPH
Quantity 2 x 30 μg Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.