missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beskrivning
Gibco™ Exosome-Depleted FBS streamlines and standardizes cell culture workflows for exosome research. It is ready to use and performance tested to help save time and enable optimal exosome preparations.
Features include:
Lowest exosome background
Enables consistent results—each lot tested for cell culture performance
No preparation needed—ultracentrifugation is not required
Exosome-Depleted FBS has been depleted of ≥90% of endogenous exosomes and is intended for use in cell culture exosome isolation assays. Every lot is tested for exosome depletion and culture performance, and our standard quality control assays are conducted to help ensure optimal and consistent results. This ready-to-use FBS for cell culture exosome isolations helps eliminate the risks associated with FBS reagent preparation by ultracentrifugation, enabling optimal assay results and increased experimental throughput.
Features include:
Exosome-Depleted FBS has been depleted of ≥90% of endogenous exosomes and is intended for use in cell culture exosome isolation assays. Every lot is tested for exosome depletion and culture performance, and our standard quality control assays are conducted to help ensure optimal and consistent results. This ready-to-use FBS for cell culture exosome isolations helps eliminate the risks associated with FBS reagent preparation by ultracentrifugation, enabling optimal assay results and increased experimental throughput.
Specifikationer
Specifikationer
| Content And Storage | –5 to –30°C |
| Description | Exosome-Depleted FBS, 100mL, Cell Culture Grade, Exosome-Depleted Serum Treatment, Store In Freezer (–5 to –30°C), Green Features Fewer Resources Used, Less Waste, For Mammalian Cell Culture |
| Country of Origin | United States |
| Format | Bottle |
| Product Type | FBS Replacement Serum |
| Quantity | 100 mL |
| Grade | Cell culture grade |
| For Use With (Application) | Mammalian cell culture |
| Purity or Quality Grade | Cell culture grade |
For Research Use Only. Not for use in diagnostic procedures.
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?