missing translation for 'onlineSavingsMsg'
Learn More

Exosome Component 9 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications

£183.00 - £395.00

Specifications

Antigen Exosome Component 9
Concentration 0.6 mg/mL
Dilution Western Blot 1:5000, Simple Western 1:1000, Chromatin Immunoprecipitation reported in scientific literature (PMID 27543448), Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported by customer review, Immunohistochemistry-Paraffin 1:50-1:100, Chromatin Immunoprecipitation (ChIP)
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18212882
View Documents
Novus Biologicals
NBP1-71702SS
0.025 mL
£183.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18701354
View Documents
Novus Biologicals
NBP1-71702
0.1 mL
£395.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Exosome Component 9 Polyclonal specifically detects Exosome Component 9 in Human, Mouse, Chicken samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

Exosome Component 9
Western Blot 1:5000, Simple Western 1:1000, Chromatin Immunoprecipitation reported in scientific literature (PMID 27543448), Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported by customer review, Immunohistochemistry-Paraffin 1:50-1:100, Chromatin Immunoprecipitation (ChIP)
Polyclonal
Rabbit
Human, Mouse, Chicken
Q06265, Q9JHI7
5393
A genomic peptide made to an internal region of human Exosome Component 9 (within residues 250-439). [Swiss-Prot Q06265]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.6 mg/mL
Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
PBS and 30% Glycerol with 0.05% Sodium Azide
Autoantigen PM/Scl 1, EC 3.1.13, exosome component 9Polymyositis/scleroderma autoantigen 75 kDa, p6, PM/Scl-75P75 polymyositis-scleroderma overlap syndrome-associated autoantigen, PMSCL175kD, Polymyositis/scleroderma autoantigen 1, polymyositis/scleroderma autoantigen 1 (75kD), polymyositis/scleroderma autoantigen 1, 75kDa
EXOSC9
IgG
Affinity Purified
Human, Mouse and Chicken. Immunogen has 83% identity to rat and 90% identity to bovine.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.